Learn More
Palladium AA Standard: Pd @ 1000 μg/mL in 20% HCl, VHG™
Product Code. 17983026
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17983026 500 mL 500mL
17933196 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17983026 Supplier VHG Labs Supplier No. VHGAAPDH500
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Palladium AA Standard: Pd @ 1000 μg/mL in 20% HCl
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Palladium
CAS 5-3-7440
Methods AA/GFAA
Analyte or Component Names Palladium
Shipping Condition Room Temperature
Molecular Formula Pd
Concentration 1000 μg/mL
Certifications/Compliance ISO 17034
Synonym Palladium on carbon, Palladium Black, Paladio, Palladium/carbon, Palladium, IG 0218A, MPP 030, MPP 050, MPP 080, P 50, P 50 (metal), Palladex 600, Palladium (nanoparticles), Palladium (powder), Palladium element, SFP 1001P, TCP 211
InChI Formula InChI=1S/Pd
SMILES [Pd]
IUPAC Name palladium
Solvent or Matrix Dilute HCl
Molecular Weight (g/mol) 106.42
Quantity 500 mL
Formula Weight 105.9035 g/mol