missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Palladin Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16179-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody, generated against N-terminal peptide of palladin, recognizes 200 kDa, 140 kDa, and 65 kDa isoforms of this protein.
Palladin is a cytoskeletal protein required for organization of normal actin cytoskeleton. Palladin plays roles in establishing cell morphology, motility, cell adhesion and cell-extracellular matrix interactions in a variety of cell types. Palladin may function as a scaffolding molecule with the potential to impact both actin polymerization as well as the assembly of existing actin filaments into higher-order arrays. It binds to proteins that bind to either monomeric or filamentous actin. It localizes at sites where active actin remodeling takes place, such as lamellipodia and membrane ruffles. Palladin′s different isoforms may have functional differences. It is also involved in the control of morphological and cytoskeletal changes associated with dendritic cell maturation.Specifications
| Palladin | |
| Polyclonal | |
| Unconjugated | |
| PALLD | |
| CGI 151, KIAA0992, palladin, PALLD, PNCA1, Sarcoma antigen NY SAR 77, SIH002 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23022 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8WX93 | |
| PALLD | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction