Learn More
Palladin Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10853-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody, generated against the fragment 999-1383 aa of palladin, recognizes most isoforms of this protein, except isoform 6.
Palladin is a cytoskeletal protein required for organization of normal actin cytoskeleton. Palladin plays roles in establishing cell morphology, motility, cell adhesion and cell-extracellular matrix interactions in a variety of cell types. Palladin may function as a scaffolding molecule with the potential to impact both actin polymerization as well as the assembly of existing actin filaments into higher-order arrays. It binds to proteins that bind to either monomeric or filamentous actin. It localizes at sites where active actin remodeling takes place, such as lamellipodia and membrane ruffles. Palladin′s different isoforms may have functional differences. It is also involved in the control of morphological and cytoskeletal changes associated with dendritic cell maturation.Specifications
| Palladin | |
| Polyclonal | |
| Unconjugated | |
| PALLD | |
| CGI 151, KIAA0992, palladin, PALLD, PNCA1, Sarcoma antigen NY SAR 77, SIH002 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23022, 72333 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8WX93, Q9ET54 | |
| Palld | |
| Palladin Fusion Protein Ag1288 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |