missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Palbociclib Sulfate Sodium Salt, TRC
Specifications
Specifications
| Chemical Name or Material | Palbociclib Sulfate Sodium Salt |
| Analyte or Component Names | Palbociclib Sulfate Sodium Salt |
| Shipping Condition | Room Temperature |
| Molecular Formula | C24 H28 N7 Na O5 S |
| InChI Formula | InChI=1S/C24H29N7O5S.Na/c1-15-19-14-26-24(28-22(19)31(17-5-3-4-6-17)23(33)21(15)16(2)32)27-20-8-7-18(13-25-20)29-9-11-30(12-10-29)37(34,35)36;/h7-8,13-14,17H,3-6,9-12H2,1-2H3,(H,34,35,36)(H,25,26,27,28);/q;+1/p-1 |
| Solution Type | Neat |
| SMILES | CC(=O)C1=C(C)c2cnc(Nc3ccc(cn3)N4CCN(CC4)S(=O)(=O)O[Na])nc2N(C5CCCC5)C1=O |
| IUPAC Name | [4-[6-[(6-acetyl-8-cyclopentyl-5-methyl-7-oxopyrido[2,3-d]pyrimidin-2-yl)amino]pyridin-3-yl]piperazin-1-yl]sulfonyloxysodium |
| Industry Type | Pharmaceutical |
| Molecular Weight (g/mol) | 549.578 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?