missing translation for 'onlineSavingsMsg'
Learn More

Palbociclib isethionate, MedChemExpress

Product Code. 16458288
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16458288 10 mg 10mg
16461976 500 mg 500mg
16450718 5 mg 5mg
16448016 100 mg 100mg
16444177 50 mg 50mg
16441356 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16458288 Supplier Medchem Express Supplier No. HYA0065

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Palbociclib (PD 0332991) isethionate is an orally active selective CDK4 and CDK6 inhibitor with IC50 values of 11 and 16 nM, respectively. Palbociclib isethionate has potent anti-proliferative activity and induces cell cycle arrest in cancer cells, which can be used in the research of HR-positive and HER2-negative breast cancer and hepatocellular carcinoma.

Palbociclib (PD 0332991) isethionate is an orally active selective CDK4 and CDK6 inhibitor with IC50 values of 11 and 16 nM, respectively. Palbociclib isethionate has potent anti-proliferative activity and induces cell cycle arrest in cancer cells, which can be used in the research of HR-positive and HER2-negative breast cancer and hepatocellular carcinoma.

Specifications

Chemical Name or Material Palbociclib isethionate
CAS 827022-33-3
Color Yellow
Purity Grade Notes Research
Molecular Formula C26H35N7O6S
Quantity 10 mg
Synonym PD 0332991 isethionate
Solubility Information H2O : 50 mg/mL (87.16 mM; Need ultrasonic) ∣DMSO : 10 mg/mL (17.43 mM; Need ultrasonic)
SMILES CC(C1=C(N2C3CCCC3)N=C(NC4=NC=C(N5CCNCC5)C=C4)N=C1)=C(C(C)=O)C2=O.OCCS(O)(=O)=O
Molecular Weight (g/mol) 573.66
Formula Weight 573.66
Percent Purity 99.52%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.