missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAK2 Rabbit anti-Human, Mouse, Rat, Clone: 2Y7I2, Novus Biologicals™
Rabbit Monoclonal Antibody
190.00 EUR - 496.00 EUR
Specifications
| Antigen | PAK2 |
|---|---|
| Clone | 2Y7I2 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18313864
|
Novus Biologicals
NBP3-16433-20UL |
20 μg |
190.00 EUR
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18331297
|
Novus Biologicals
NBP3-16433-100UL |
100 μg |
496.00 EUR
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PAK2 Monoclonal antibody specifically detects PAK2 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
| PAK2 | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Protein Kinase | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 5062 | |
| IgG | |
| Affinity purified |
| 2Y7I2 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| EC 2.7.11, gamma-PAK, p21 (CDKN1A)-activated kinase 2, p21 protein (Cdc42/Rac)-activated kinase 2, p21-activated kinase 2, p58, PAK-2, PAK65EC 2.7.11.1, PAKgamma, S6/H4 kinase, serine/threonine-protein kinase PAK 2 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human PAK2 (Q13177). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title