Learn More
Padsevonil, MedChemExpress
Product Code. 16473006
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. 16473006
Brand: Medchem Express HY109009
MedChemExpress Padsevonil (UCB0942) is a first-in-class and potent antiepileptic agent. Padsevonil can be used for the research of epilepsy.
Padsevonil (UCB0942) is a first-in-class and potent antiepileptic agent. Padsevonil can be used for the research of epilepsy.
Specifications
Chemical Name or Material Padsevonil
CAS 1294000-61-5
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C14H14ClF5N4O2S
Quantity 25 mg
Synonym UCB-0942
Solubility Information DMSO : 100 mg/mL (231.05 mM; Need ultrasonic)
SMILES O=C1N(C[C@@H](CC(F)(F)Cl)C1)CC(N(N=C(COC)S2)C2=N3)=C3C(F)(F)F
Molecular Weight (g/mol) 432.8
Formula Weight 432.8
Percent Purity 97.42%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation