Learn More
Kern Sohn Padded Plastic Box for Custom Weight Set
Description
- Plastic
- For button/compact weight design
Specifications
Specifications
| Product Type | Case |
| Material | Plastic |
| For Use With (Equipment) | Class E1 and M3 Weight |
| Holds | 1 mg to 500 g weights |