Learn More
PABPC1 PABP Mouse anti-Human, Unconjugated, Clone: 3D8B5, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66809-1-IG-20UL
 View more versions of this product
Product Code. 16869025
  • 145.00 EUR / 20µL
Description
The PABPC1, PABP antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PABPC1, PABP. This antibody recognizes human, rat antigen. The PABPC1, PABP antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
PABPC1 PABP
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
3D8B5
Unconjugated
20 μL
26986
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions