missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PA2G4 Mouse anti-Human, Mouse, Rat, Clone: 3G10D9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66055-1-IG-150UL
This item is not returnable.
View return policy
Description
This antibody is a mouse mono-clonal antibody raised against full length PA2G4 of human origin.
This gene encodes an RNA-binding protein that is involved in growth regulation. This protein is present in pre-ribosomal ribonucleoprotein complexes and may be involved in ribosome assembly and the regulation of intermediate and late steps of rRNA processing. This protein can interact with the cytoplasmic domain of the ErbB3 receptor and may contribute to transducing growth regulatory signals. This protein is also a transcriptional co-repressor of androgen receptor-regulated genes and other cell cycle regulatory genes through its interactions with histone deacetylases. This protein has been implicated in growth inhibition and the induction of differentiation of human cancer cells. Six pseudogenes, located on chromosomes 3, 6, 9, 18, 20 and X, have been identified.Specifications
| PA2G4 | |
| Monoclonal | |
| 1.04 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P50580, Q6AYD3, Q9UQ80 | |
| Pa2g4 | |
| PA2G4 Fusion Protein Ag7850 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2a |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 3G10D9 | |
| Unconjugated | |
| PA2G4 | |
| EBP1, ErbB3 binding protein 1, HG4 1, p38 2G4, PA2G4 | |
| Mouse | |
| Protein A | |
| RUO | |
| 18813, 288778, 5036 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction