Learn More
p65/RELA Rabbit anti-Human, Mouse, Porcine, Rat, Polyclonal, Proteintech
Product Code. 16817333 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16817333 20 μL 20µL
16807333 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16817333 Supplier Proteintech Supplier No. 107451AP20UL
Rabbit Polyclonal Antibody
This antibody is a rabbit polyclonal antibody raised against residues near the N terminus of human RELA.
The transcription factor NFkB is widely recognized as a critical mediator of immune and inflammatory responses. In most cell types, NFkB is found in the cytoplasm where it is associated with an inhibitory protein known as IkB. An impressive variety of stimuli (tumor necrosis factor, interleukin 1, T-cell activation signals, bacterial endotoxins, viral transforming proteins, certain growth factors and reactive oxygen intermediates) lead to the rapid nuclear assumulation of NFkB by the induced phosphorylation and subsquent degradation of IkB. In the nucleus, NFkB regulates genes encoding cytokines, cytokine receptors, cell adhesion molecules, proteins involved in coagulation and genes involved in cell growth control. Additionally, NFkB is thought to be an important transcriptional regulator for HIV. Growing evidence indicates that the dysregulation of NFkB may be key to a number of diseases including arthritis and other inflammatory diseases, Alzheimer′s disease, atherosclerosis, and cancer.
Specifications
Antigen p65/RELA
Applications Flow Cytometry, Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.52 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene RELA
Gene Accession No. Q04206, Q04207
Gene Alias NFKB 3, NFKB p65, NFKB3, p65, RELA, Transcription factor p65
Gene Symbols RELA
Host Species Rabbit
Immunogen p65; RELA Fusion Protein Ag1199
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100135665, 19697, 309165, 5970
Target Species Human, Pig, Rat, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG