missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ p53 [ac Lys382] Antibody (SR2907), Novus Biologicals™
Rabbit Monoclonal Antibody
316.00 EUR - 382.00 EUR
Specifications
| Antigen | p53 |
|---|---|
| Clone | SR2907 |
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:100 |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30041803
|
Novus Biologicals™
NBP3-27721-100ul |
100 μL |
382.00 EUR
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30041688
|
Novus Biologicals™
NBP3-27721-50ul |
50 μL |
316.00 EUR
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
p53 Monoclonal antibody specifically detects p53 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunocytochemistry/ ImmunofluorescenceSpecifications
| p53 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Antigen NY-CO-13, FLJ92943, LFS1TRP53, p53, p53 tumor suppressor, P53cellular tumor antigen p53, Phosphoprotein p53, transformation-related protein 53, tumor protein p53, Tumor suppressor p53 | |
| A synthesized peptide derived from human p53 (Uniprot #: P04637) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| SR2907 | |
| Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cancer, Cell Cycle and Replication, Cellular Markers, Checkpoint signaling, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, HIF Target Genes, Hypoxia, Neuroscience, Neurotransmission, p53 Pathway, Phospho Specific, Stem Cell Markers, Transcription Factors and Regulators, Tumor Suppressors | |
| PBS (pH 7.4), 150mM NaCl, 50% glycerol | |
| 7157 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title