missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p40/deltaNp63 Rabbit anti-Human, Clone: P40/4396R, Novus Biologicals™
Rabbit Monoclonal Antibody
306.00 EUR - 551.00 EUR
Specifications
| Antigen | p40/deltaNp63 |
|---|---|
| Clone | P40/4396R |
| Concentration | 0.2 mg/ml |
| Dilution | Flow Cytometry 1-2 ug/million cells, ELISA, Immunohistochemistry-Paraffin 1-2 ug/ml |
| Applications | Flow Cytometry, ELISA, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18331617
|
Novus Biologicals
NBP3-13805-20UG |
20 μg |
306.00 EUR
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18392134
|
Novus Biologicals
NBP3-13805-100UG |
100 μg |
551.00 EUR
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
p40/deltaNp63 Monoclonal antibody specifically detects p40/deltaNp63 in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry (Paraffin)Specifications
| p40/deltaNp63 | |
| 0.2 mg/ml | |
| Flow Cytometry, ELISA, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q9H3D4 | |
| 8626 | |
| IgG κ | |
| Protein A purified |
| P40/4396R | |
| Flow Cytometry 1-2 ug/million cells, ELISA, Immunohistochemistry-Paraffin 1-2 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| AIS, amplified in squamous cell carcinoma, B(p51A), B(p51B), Chronic ulcerative stomatitis protein, EEC3, Keratinocyte transcription factor KET, KET, LMS, OFC8, p53CP, SHFM4, TP53CP, TP53L, TP63, TP73L, Transformation-related protein 63, tumor protein p53-competing protein | |
| Recombinant fragment (around aa1-100) of human p40/deltaNp63 protein (exact sequence is proprietary) (Uniprot: Q9H3D4 ) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title