missing translation for 'onlineSavingsMsg'
Learn More

p38 MAPK-IN-1, MedChemExpress

Product Code. p-200101955
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16475388 10 mg 10mg
16461577 50 mg 50mg
16438107 5 mg 5mg
16416006 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16475388 Supplier Medchem Express Supplier No. HY12839

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress p38 MAPK-IN-1 (Compound 4) is a novel potent and selective inhibitor of p38 MAPK with IC50 of 68 nM. p38 MAPK-IN-1 shows sustained levels, low clearance and good bioavailability.

p38 MAPK-IN-1 (Compound 4) is a novel potent and selective inhibitor of p38 MAPK with IC50 of 68 nM. p38 MAPK-IN-1 shows sustained levels, low clearance and good bioavailability.

Specifications

Chemical Name or Material p38 MAPK-IN-1
CAS 1006378-90-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C21H15F2N2O
Quantity 10 mg
Solubility Information DMSO : 8.33 mg/mL (23.84 mM; Need ultrasonic)
SMILES FC1=CC(F)=C(C2=CC=NC(C2=CC=[NH+]3[O-])=C3C4=C(C)C=CC=C4)C=C1
Molecular Weight (g/mol) 349.35
Formula Weight 349.35
Percent Purity 98.09%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.