Learn More
p38 MAPK-IN-1, MedChemExpress
Product Code. 16461577
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16461577
Brand: Medchem Express HY12839
MedChemExpress p38 MAPK-IN-1 (Compound 4) is a novel potent and selective inhibitor of p38 MAPK with IC50 of 68 nM. p38 MAPK-IN-1 shows sustained levels, low clearance and good bioavailability.
p38 MAPK-IN-1 (Compound 4) is a novel potent and selective inhibitor of p38 MAPK with IC50 of 68 nM. p38 MAPK-IN-1 shows sustained levels, low clearance and good bioavailability.
Specifications
Chemical Name or Material p38 MAPK-IN-1
CAS 1006378-90-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C21H15F2N2O
Quantity 50 mg
Solubility Information DMSO : 8.33 mg/mL (23.84 mM; Need ultrasonic)
SMILES FC1=CC(F)=C(C2=CC=NC(C2=CC=[NH+]3[O-])=C3C4=C(C)C=CC=C4)C=C1
Molecular Weight (g/mol) 349.35
Formula Weight 349.35
Percent Purity 98.09%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease