missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p38 MAPK-IN-1, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
p38 MAPK-IN-1 (Compound 4) is a novel potent and selective inhibitor of p38 MAPK with IC50 of 68 nM. p38 MAPK-IN-1 shows sustained levels, low clearance and good bioavailability.
Specifications
Specifications
| Chemical Name or Material | p38 MAPK-IN-1 |
| CAS | 1006378-90-0 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C21H15F2N2O |
| Quantity | 5 mg |
| Solubility Information | DMSO : 8.33 mg/mL (23.84 mM; Need ultrasonic) |
| SMILES | FC1=CC(F)=C(C2=CC=NC(C2=CC=[NH+]3[O-])=C3C4=C(C)C=CC=C4)C=C1 |
| Molecular Weight (g/mol) | 349.35 |
| Formula Weight | 349.35 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction