missing translation for 'onlineSavingsMsg'
Learn More

p38 MAPK-IN-1, MedChemExpress

Product Code. 16416006
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16416006

Brand: Medchem Express HY12839

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress p38 MAPK-IN-1 (Compound 4) is a novel potent and selective inhibitor of p38 MAPK with IC50 of 68 nM. p38 MAPK-IN-1 shows sustained levels, low clearance and good bioavailability.

p38 MAPK-IN-1 (Compound 4) is a novel potent and selective inhibitor of p38 MAPK with IC50 of 68 nM. p38 MAPK-IN-1 shows sustained levels, low clearance and good bioavailability.

Specifications

Chemical Name or Material p38 MAPK-IN-1
CAS 1006378-90-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C21H15F2N2O
Quantity 100 mg
Solubility Information DMSO : 8.33 mg/mL (23.84 mM; Need ultrasonic)
SMILES FC1=CC(F)=C(C2=CC=NC(C2=CC=[NH+]3[O-])=C3C4=C(C)C=CC=C4)C=C1
Molecular Weight (g/mol) 349.35
Formula Weight 349.35
Percent Purity 98.09%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.