missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ P3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554486
This item is not returnable.
View return policy
Description
Immunogen sequence: DSEGIIVISS QYPGQANRTA PGPMLRVTSL DTEVLTIKNV SAITWGGGGG FVVSIHSGLA GLAPLHIQLV DAHEAPPTLI EERRDFCIKV SPAEDTPATL SADLAHFSEN PILYLLLPLI FVNKCSFGCK VELEVLKGLM Q Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 84%.
The ubiquitous expression and the conservation of the sequence in distant animal species suggest that the gene codes for a protein with housekeeping functions.
Specifications
| P3 | |
| Polyclonal | |
| Unconjugated | |
| SLC10A3 | |
| DXHXS253E; DXS253E; DXS253Eh; DXSmhG28; P3; P3 protein; Protein P3; SLC10A3; solute carrier family 10 (sodium/bile acid cotransporter family), member 3; solute carrier family 10 member 3; solute carrier family 10, member 3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8273 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P09131 | |
| SLC10A3 | |
| Recombinant protein corresponding to Human P3. Recombinant protein control fragment (Product #RP-92200). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction