missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ P2Y11 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16672502
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16672502 100 μL 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16672502 Supplier Invitrogen™ Supplier No. PA5102684

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total P2RY11.

P2Y11 belongs to the family of G protein coupled receptors, and is coupled to the stimulation of the phosphoinositide and adenylyl cyclase pathways and behaves as a selective purinoceptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen P2Y11
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene P2RY11
Gene Accession No. Q96G91
Gene Alias P2RY11; P2Y purinoceptor 11; P2Y11; purinergic receptor P2Y, G-protein coupled, 11; purinergic receptor P2Y11
Gene Symbols P2RY11
Host Species Rabbit
Immunogen A synthesized peptide derived from human P2RY11(Accession Q96G91), corresponding to amino acid residues L312-P362.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5032
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.