Learn More
Lonza™ P1 Primary Cell 4D-Nucleofector™ X Kit S
Description
- High transfection efficiencies for primary cells.
- Cells maintain functionality post transfection.
- Conditions are transferable between 4D-Nucleofector™ System, 96-well Shuttle™ System and 384-well Nucleofector™ System.
- Applications:
- Transfection of cell numbers in the range of 2 x 105 to 2 x 107 cells.
- Low throughput applications.
- This kit has been determined to be optimal for transfecting following cell types:
- Macrophage (mouse)
- NHDF (human), neonatal
Specifications
Specifications
| Color | Clear |
| Vector | pmaxGFP™ |
| For Use With (Equipment) | 4D-Nucleofector™ X Unit (20 μL format) |
| For Use With (Application) | Efficient transfection of specific primary cells, e.g., human ADSCs or MSCs |
| Includes | 2 x 0.675 mL 1 Primary Cell Nucleofector™ Solution, 2 x 0.15 mL Supplement 1, 50 μg pmaxGFP™ Vector (1 μg/μL in 10 mM Tris pH 8.0), 2 x 16-well Nucleocuvette™ Strips (20 μL) |
| No. of Reactions | 32 |
| pH | 7.4 to 7.6 |
| Quantity | 1 Kit |