missing translation for 'onlineSavingsMsg'
Learn More

p-Sulfamylacetanilide, TRC

Product Code. 30137945
Change view
Click to view available options
Quantity:
500 mg
5 g
10 g
Unit Size:
10g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137945 5 g 5g
30138311 10 g 10g
30006771 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137945 Supplier Toronto Research Chemicals Supplier No. TRCS6992405G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-61-9
Molecular Formula C8 H10 N2 O3 S
Molecular Weight (g/mol) 214.24
Synonym Acetamide, N-[4-(aminosulfonyl)phenyl]-, Acetanilide, 4'-sulfamoyl- (6CI,7CI,8CI), 4-Acetamidobenzenesulfonamide, 4-Acetylaminobenzenesulfonamide, 4'-Sulfamoylacetanilide, A-319, Acetylsulfanilamide, Benzenesulfonamide, 4-(acetylamino)-, Erytrin, N-(4-Sulfamoylphenyl)acetamide, N-[4-(Aminosulfonyl)phenyl]acetamide, N4-Acetsulfanilamide, N4-Acetylsulfanilamide, NSC 217, NSC 406839, Neotherapol, N'-Acetylsulphanilamide, Sulfanilamide-N4-acetate, p-(Acetylamino)benzenesulfonamide, p-Acetamidobenzenesulfonamide, p-Sulfamoylacetanilide, p-Sulfamylacetanilide, Sulfanilamide-N-acetyl
IUPAC Name N-(4-sulfamoylphenyl)acetamide
SMILES CC(=O)Nc1ccc(cc1)S(=O)(=O)N

Specifications

Quantity 5 g
InChI Formula InChI=1S/C8H10N2O3S/c1-6(11)10-7-2-4-8(5-3-7)14(9,12)13/h2-5H,1H3,(H,10,11)(H2,9,12,13)
Formula Weight 214.0412
Chemical Name or Material Sulfanilamide-N-acetyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.