Learn More
Welch™ P 6 Z Chemvac Combination Pump System
Shop All Welch Ilmvac Products
Click to view available options
Depth (Metric):
330 mm
Unit Size:
Each
Specifications
Specifications
| Electrical Requirements | 230 V 50/60 Hz |
| Depth (Metric) | 330 mm |
| Model | P 6 Z |
| Flow Rate | 97/110 L/min. |
| Frequency | 50/60 Hz |
| Overall Height (Metric) | 340 mm |
| Overall Width (Metric) | 500 mm |
| Pump Type | Chemvac Combination Pump System |
| Maximum Pressure | 2 x 10-3 mbar |
| Voltage | 230 V |