Learn More
Welch™ P 23 Z - 301 Chemvac Combination Pump System
Shop All Welch Ilmvac Products
Click to view available options
Depth (Metric):
345 mm
Unit Size:
Each
Specifications
Specifications
| Electrical Requirements | 115/230 V 50/60 Hz |
| Depth (Metric) | 345 mm |
| Model | P 23 Z - 301 |
| Frequency | 50/60 Hz |
| Overall Height (Metric) | 420 mm |
| Overall Width (Metric) | 590 mm |
| Pump Type | Chemvac Combination Pump System |
| Maximum Pressure | 2 x 10-3 mbar |
| Voltage | 115/230 V |