Learn More
Welch™ Chemvac P12Z - 301 Combination Pump
Shop All Welch Ilmvac Products
Click to view available options
Depth (Metric):
345 mm
Unit Size:
Each
Description
- Comprised of two-stage rotary vane pump P 6 Z and two-stage diaphragm pump MPC 301 Z
- Suitable for freeze dry applications
- Both pumps are electrical and vacuum-connected
- Rotary vane pump includes an outtake with a separator for the collection of condensates.
- Supplied completed with solenoid valve block
Specifications
Specifications
| Electrical Requirements | 115/230 V 50/60 Hz |
| Depth (Metric) | 345 mm |
| Model | P 12 Z - 301 |
| Frequency | 50/60 Hz |
| Overall Height (Metric) | 420 mm |
| Overall Width (Metric) | 590 mm |
| Pump Type | Chemvac Combination Pump System |
| Maximum Pressure | 2 x 10-3 mbar |
| Voltage | 115/230 V |
Product Content Correction
Product Title
Ser du en mulighed for forbedring?Del en indholdskorrektion