missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oxymetholone, 1mg/ml in Acetonitrile, Lipomed
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Description
Oxymetholone, 1mg/ml in Acetonitrile
Specifications
Specifications
| Chemical Name or Material | Oxymetholone |
| CAS | 434-07-1 |
| Analyte or Component Names | Oxymetholone |
| Shipping Condition | Ice Pk. (-20°C) |
| Molecular Formula | C21 H32 O3 |
| Concentration | 1 mg/mL |
| Synonym | 17-Beta-Hydroxy-2-hydroxymethylene-17-alpha-methyl-3-androstanone, 2-(Hydroxymethylene)-17-methyldihydrotestosterone, Adroyd, Anadrol, Anapolan 50, 2-Hydroxymethylene-17 beta-hydroxy-17 alpha-methyl-5 alpha-androstan-3-one, 2-Hydroxymethylene-17 beta-hydroxy-17 alpha-methyl-5 alpha-androstan-3-one, 2-Hydroxymethylene-17 beta-hydroxy-17 alpha-methyl-5 alpha-androstan-3-one, 2-Hydroxymethylene-17 beta-hydroxy-17 alpha-methyl-5 alpha-androstan-3-one, 2-Hydroxymethylene-17 beta-hydroxy-17 alpha-methyl-5 alpha-androstan-3-one, 2-Hydroxymethylene-17 beta-hydroxy-17 alpha-methyl-5 alpha-androstan-3-one, 2-Hydroxymethylene-17 beta-hydroxy-17 alpha-methyl-5 alpha-androstan-3-one, 2-Hydroxymethylene-17 beta-hydroxy-17 alpha-methyl-5 alpha-androstan-3-one, 2-Hydroxymethylene-17 beta-hydroxy-17 alpha-methyl-5 alpha-androstan-3-one, 2-Hydroxymethylene-17 beta-hydroxy-17 alpha-methyl-5 alpha-androstan-3-one, 2-Hydroxymethylene-17 beta-hydroxy-17 alpha-methyl-5 alpha-androstan-3-one, 2-Hydroxymethylene-17 beta-hydroxy-17 alpha-methyl-5 alpha-androstan-3-one, 2-Hydroxymethylene-17 beta-hydroxy-17 alpha-methyl-5 alpha-androstan-3-one, 2-Hydroxymethylene-17 beta-hydroxy-17 alpha-methyl-5 alpha-androstan-3-one, 2-Hydroxymethylene-17 beta-hydroxy-17 alpha-methyl-5 alpha-androstan-3-one, 2-Hydroxymethylene-17 beta-hydroxy-17 alpha-methyl-5 alpha-androstan-3-one, 2-Hydroxymethylene-17 beta-hydroxy-17 alpha-methyl-5 alpha-androstan-3-one, 2-Hydroxymethylene-17 beta-hydroxy-17 alpha-methyl-5 alpha-androstan-3-one, 2-Hydroxymethylene-17 beta-hydroxy-17 alpha-methyl-5 alpha-androstan-3-one, 2-Hydroxymethylene-17 beta-hydroxy-17 alpha-methyl-5 alpha-androstan-3-one, 2-Hydroxymethylene-17 beta-hydroxy-17 alpha-methyl-5 alpha-androstan-3-one, 2-Hydroxymethylene-17 beta-hydroxy-17 alpha-methyl-5 alpha-androstan-3-one, 2-Hydroxymethylene-17 beta-hydroxy-17 alpha-methyl-5 alpha-androstan-3-one, 2-Hydroxymethylene-17 beta-hydroxy-17 alpha-methyl-5 alpha-androstan-3-one, 2-Hydroxymethylene-17 beta-hydroxy-17 alpha-methyl-5 alpha-androstan-3-one, 2-Hydroxymethylene-17 beta-hydroxy-17 alpha-methyl-5 alpha-androstan-3-one |
| InChI Formula | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMILES | C[C@]1(O)CC[C@H]2 [C@@H]3 CC[C@H]4 CC(=O)\C(=C/O)\C[C@]4(C)[C@H]3 CC[C@]12 C |
| IUPAC Name | (2 Z,5 S,8 R,9 S,10 S,13 S,14 S,17 S)-17-hydroxy-2-(hydroxymethylidene)-10,13,17-trimethyl-1,4,5,6,7,8,9,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthren-3-one |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction