Learn More
(±)-Oxybutynin-d11 HCl (cyclohexyl-d11), CDN
Specifications
Specifications
| Chemical Name or Material | Oxybutynin-d11 Chloride |
| CAS | 1185151-95-4 |
| Molecular Formula | C22 D11 H20 N O3 . Cl H |
| Quantity | 0.05 g |
| Synonym | 4-Diethylaminobut-2-ynyl 2-hydroxy-2-phenyl-2-(1,2,2,3,3,4,4,5,5,6,6-undecadeuteriocyclohexyl)acetate hydrochloride, 4-(Diethylamino)-2-butyn-1-yl α-(cyclohexyl-d11)-α-hydroxybenzeneacetate hydrochloride, (±)-Oxybutynin-d11 HCl (cyclohexyl-d11), Oxybutynin-d11 Chloride, Oxybutynin-D11 (cyclohexyl-D11) Hydrochloride |
| InChI Formula | InChI=1 S/C22H31NO3.ClH/c1-3-23(4-2)17-11-12-18-26-21(24)22(25,19-13-7-5-8-14-19)20-15-9-6-10-16-20;/h5,7-8,13-14,20,25 H,3-4,6,9-10,15-18H2,1-2H3;1 H/i6D2,9D2,10D2,15D2,16D2,20 D; |
| SMILES | Cl.[2 H]C1([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])(C(O)(C(=O)OCC#CCN(CC)CC)c2ccccc2)C([2 H])([2 H])C1([2 H])[2 H] |
| IUPAC Name | 4-(diethylamino)but-2-ynyl 2-hydroxy-2-phenyl-2-(1,2,2,3,3,4,4,5,5,6,6-undecadeuteriocyclohexyl)acetate;hydrochloride |
| Molecular Weight (g/mol) | 405.015 |
| Formula Weight | 404.276 g/mol |
| Show More |