missing translation for 'onlineSavingsMsg'
Learn More

Oxybutynin-d11 Chloride, TRC

Product Code. 30179382
Change view
Click to view available options
Quantity:
1 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179382 10 mg 10mg
30179598 25 mg 25mg
30179653 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30179382 Supplier Toronto Research Chemicals Supplier No. TRCO86852710MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1185151-95-4
Molecular Formula C22 D11 H20 N O3 . Cl H
Molecular Weight (g/mol) 405.015
Synonym 4-Diethylaminobut-2-ynyl 2-hydroxy-2-phenyl-2-(1,2,2,3,3,4,4,5,5,6,6-undecadeuteriocyclohexyl)acetate hydrochloride, 4-(Diethylamino)-2-butyn-1-yl α-(cyclohexyl-d11)-α-hydroxybenzeneacetate hydrochloride, (±)-Oxybutynin-d11 HCl (cyclohexyl-d11), Oxybutynin-d11 Chloride, Oxybutynin-D11 (cyclohexyl-D11) Hydrochloride
IUPAC Name 4-(diethylamino)but-2-ynyl 2-hydroxy-2-phenyl-2-(1,2,2,3,3,4,4,5,5,6,6-undecadeuteriocyclohexyl)acetate;hydrochloride
SMILES Cl.[2H]C1([2H])C([2H])([2H])C([2H])([2H])C([2H])(C(O)(C(=O)OCC#CCN(CC)CC)c2ccccc2)C([2H])([2H])C1([2H])[2H]

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C22H31NO3.ClH/c1-3-23(4-2)17-11-12-18-26-21(24)22(25,19-13-7-5-8-14-19)20-15-9-6-10-16-20;/h5,7-8,13-14,20,25H,3-4,6,9-10,15-18H2,1-2H3;1H/i6D2,9D2,10D2,15D2,16D2,20D;
Formula Weight 404.276
Chemical Name or Material Oxybutynin-d11 Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.