Learn More
Oxybutynin-d11 Chloride, TRC
Product Code. 30179598
Change view
Click to view available options
Quantity:
1 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179598 25 mg 25mg
30179653 1 mg 1mg
30179382 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30179598 Supplier Toronto Research Chemicals Supplier No. TRCO86852725MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1185151-95-4
Molecular Formula C22 D11 H20 N O3 . Cl H
Molecular Weight (g/mol) 405.015
Synonym 4-Diethylaminobut-2-ynyl 2-hydroxy-2-phenyl-2-(1,2,2,3,3,4,4,5,5,6,6-undecadeuteriocyclohexyl)acetate hydrochloride, 4-(Diethylamino)-2-butyn-1-yl α-(cyclohexyl-d11)-α-hydroxybenzeneacetate hydrochloride, (±)-Oxybutynin-d11 HCl (cyclohexyl-d11), Oxybutynin-d11 Chloride, Oxybutynin-D11 (cyclohexyl-D11) Hydrochloride
IUPAC Name 4-(diethylamino)but-2-ynyl 2-hydroxy-2-phenyl-2-(1,2,2,3,3,4,4,5,5,6,6-undecadeuteriocyclohexyl)acetate;hydrochloride
SMILES Cl.[2H]C1([2H])C([2H])([2H])C([2H])([2H])C([2H])(C(O)(C(=O)OCC#CCN(CC)CC)c2ccccc2)C([2H])([2H])C1([2H])[2H]
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C22H31NO3.ClH/c1-3-23(4-2)17-11-12-18-26-21(24)22(25,19-13-7-5-8-14-19)20-15-9-6-10-16-20;/h5,7-8,13-14,20,25H,3-4,6,9-10,15-18H2,1-2H3;1H/i6D2,9D2,10D2,15D2,16D2,20D;
Formula Weight 404.276
Chemical Name or Material Oxybutynin-d11 Chloride