Learn More
Thermo Scientific™ Oxoid™ Sodium Chloride Bacteriological
Click to view available options
Quantity:
500 g
Unit Size:
500g
Description
Oxoid Sodium Chloride Bacteriological is a dehydrated salt suitable for inclusion in bacteriological culture media.
Specifications
Specifications
| Description | Dehydrated; Sodium chloride bacteriological |
| Form | Powder |
| Product Type | Sodium Chloride |
| Quantity | 500 g |