Learn More
Thermo Scientific™ Oxoid™ Peptonised Milk
Product Code. 10706593
Click to view available options
Quantity:
250g
Unit Size:
250g
Product Code. 10706593
Brand: Thermo Scientific™ LP0032J
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Oxoid Peptonised Milk is a pancreatic digest of high grade skimmed milk powder with high levels of carbohydrate and calcium.
Oxoid Peptonised Milk is a pancreatic digest of high grade skimmed milk powder with high levels of carbohydrate and calcium.
  • Particularly useful in media for bacteriological examination of dairy products
TRUSTED_SUSTAINABILITY
Specifications
Description Peptonized milk
Form Powder
Product Type Milk
Quantity 250g