Learn More
Thermo Scientific™ Oxoid™ Peptonised Milk
Click to view available options
Quantity:
250g
Unit Size:
250g
Description
Oxoid Peptonised Milk is a pancreatic digest of high grade skimmed milk powder with high levels of carbohydrate and calcium.
- Particularly useful in media for bacteriological examination of dairy products
Specifications
Specifications
| Description | Peptonized milk |
| Form | Powder |
| Product Type | Milk |
| Quantity | 250g |