missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Modified Edwards Medium (Dehydrated)

Product Code. 10699924
Click to view available options
Quantity:
500 g
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 10699924

Brand: Thermo Scientific™ CM0027B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rapidly isolate Streptococcus agalactiae and other streptococci involved in bovine mastitis with this selective medium.

Rapidly isolate Streptococcus agalactiae and other streptococci involved in bovine mastitis with selective Thermo Scientific™ Oxoid™ Modified Edwards Medium (Dehydrated). Aesculin in the medium differentiates the negative Streptococcus agalactiae (blue colonies) from aesculin-positive Group D streptococci (black colonies). Crystal violet and thallus sulphate serve as the selective agents for the streptococci.

Streptococci are gram-positive facultatively anaerobic bacteria, which constitute normal commensal flora of mouth, skin, intestine and upper respiratory tract of humans. The Strep organisms that cause mastitis are commonly divided into Streptococcus agalactiae and Streps other than Streptococcus agalactiae. Streptococcus agalactiae is a major cause of subclinical mastitis in dairy cattle and a source of economic loss for the industry.

Use Modified Edwards Medium for rapid isolation of Streptococcus agalactiae and other streptococci involved in bovine mastitis.

  • Selective: crystal violet and thallus sulphate serve as the selective agents for the streptococci.
  • Easy-to-read: Streptococcus agalactiae (blue colonies) are easy to differentiate from aesculin-positive Group D streptococci (black colonies).

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description Edwards Medium, Modified
Form Powder
Product Type Dehydrated Culture Media
Quantity 500 g
Yield For 9.6L medium

For Laboratory Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.