Learn More
Thermo Scientific™ Oxoid™ Legionella BCYE Supplement w/o Cysteine
Oxoid Legionella BCYE Supplement w/o L-cysteine is used for the isolation of Legionella spp.
Brand:  Thermo Scientific™ SR0175A
Product Code. 11973242
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Oxoid Legionella BCYE Supplement w/o L-cysteine is used for the isolation of Legionella spp.
  • Add to Legionella CYE Agar Base, Part No. CM1203B
  • Each vial supplements 100mL of medium
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Legionella BCYE Supplement w/o L-cysteine
Supplement
10/Pk.
Each vial for 100mL medium