Learn More
Thermo Scientific™ Oxoid™ Legionella BCYE Supplement w/o Cysteine
Oxoid Legionella BCYE Supplement w/o L-cysteine is used for the isolation of Legionella spp.
Brand: Thermo Scientific™ SR0175A
This item is not returnable.
View return policy
Description
Oxoid Legionella BCYE Supplement w/o L-cysteine is used for the isolation of Legionella spp.
- Add to Legionella CYE Agar Base, Part No. CM1203B
- Each vial supplements 100mL of medium
Specifications
| Legionella BCYE Supplement w/o L-cysteine | |
| Supplement | |
| 10/Pk. | |
| Each vial for 100mL medium |