Learn More
Thermo Scientific™ Hynes Desoxycholate Citrate Agar (Dehydrated)
Description
Isolate and differentiate Salmonella and Shigella species from clinical samples easily and with improved selectivity using Thermo Scientific™ Oxoid™ Hynes Desoxycholate Citrate Agar (Dehydrated).
The medium is pale pink and clear. Lactose fermenting organisms produce pink colonies and may be surrounded by a zone of precipitated desoxycholic acid. The colonies of non-lactose fermenters are colorless, and, due to their alkaline reaction, they are surrounded by a clear orange-yellow zone of medium.
An improved medium, based on the Hynes1 modification of Leifsons2 medium, Desoxycholate Citrate Agar (Hynes Modification) makes the isolation of salmonellae and shigellae easier.
- Easier colony selection: improved medium gives larger and more numerous colonies of Shigella species which can easily be picked off and emulsified in saline for slide agglutination tests
- Improved selectivity: more inhibitory to coliforms and Proteus species than the original Desoxycholate Citrate Agar (Part No. CM0035B)
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Specifications
| Description | Desoxycholate Citrate Agar, Hynes |
| Form | Powder |
| Product Type | Agar |
| Quantity | 500 g |
| Yield | For 9.6L medium |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.