Learn More
Thermo Scientific™ Antibiotic Medium No. 1 (Dehydrated)
Product Code. 10649364
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10649364 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10649364 Supplier Thermo Scientific™ Supplier No. CM0327B
Carry out microbiological assay of antibiotics with the seed layer of Antibiotic Medium No.1
Carry out microbiological assay of antibiotics with the seed layer of Thermo Scientific™ Oxoid™ Antibiotic Medium No.1 (Dehydrated). This is one of the most important media in antibiotic assay work and it is in a specially modified form to take advantage of the properties of Oxoid Agar No.1. (LP0011).
Certain agars have inhibitory properties towards some antibiotics, particularly streptomycin, kanamycin, polymyxin B and neomycin1. Oxoid Antibiotic Medium No.1 does not share these inhibitory properties and is therefore especially suited for antibiotic assay work.
Antibiotic Medium No.1 is a recommended medium for the seed layer in the preparation of plates for the microbiological assay of antibiotics.
  • The medium is used as a seed agar with Micrococcus flavus for the plate assay of bacitracin; with Sarcina lutea for the plate assay of chloramphenicol and with Staphylococcus aureus for the assay of kanamycin sulphate, penicillin G, sodium methicillin and sodium oxacillin.
  • Employed as a base agar in the assay of the number of drugs like chloramphenicol, kanamycin sulphate, colistin sulphate, sodium methicillin, sodium oxacillin and vancomycin hydrochloride.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Industrial Reference: AOAC, FDA, USDA, ISO.
Description Antibiotic Medium No. 1
Form Powder
Product Type Dehydrated Culture Media
Quantity 500 g
Yield For 18.5L medium