Learn More
WTW™ OxiTop™-i Respirometric BOD Measuring System
Click to view available options
Type:
OxiTop™-IDS 1
OxiTop™-IDS 12
OxiTop™-i 2
OxiTop™-i 6
OxiTop™-i B
OxiTop™-i IS 12
OxiTop™-i IS 6
OxiTop™-i SET 2
Unit Size:
1 set
Each
Pack of 12
Description
- OxiTop™ is a respirometric vessel-top measuring system for BOD self-check measurement with undiluted samples (DIN EN 1899 -2)
- Measuring ranges up to 4000 mg/L BOD possible
- No need to convert data - direct input of sample volume
- Direct display of the BOD value on the measuring head screen
- Menu-operated, with backlit graphic LCD, control LED and operating buttons for direct BOD readings, volume pre-selection, measurement duration between 1 and 7 days selectable, display of measuring curve, display of current readings
Spécification
Spécification
| Type | OxiTop™-i IS 6 |
| Display Type | Backlit Graphic LCD |
| Range | Measuring ranges up to 4000 mg/L BOD possible |
| Includes | Six OxiTop™-i measuring heads, universal plug-in power supply 100-240 VAC, 50/60 Hz |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu