missing translation for 'onlineSavingsMsg'
Learn More

Oxiracetam, TRC

Product Code. 30138591
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138591 25 mg 25mg
30133560 100 mg 100mg
17983415 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138591 Supplier Toronto Research Chemicals Supplier No. TRCO84690525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62613-82-5
Molecular Formula C6 H10 N2 O3
Molecular Weight (g/mol) 158.16
Synonym 2-(4-Hydroxy-2-oxopyrrolidin-1-yl)acetamide, 4-Hydroxy-2-oxo-1-pyrrolidineacetamide, 4-Hydroxypyrrolidone-2-acetamide, CGP 21690, CGP 21690E, CT 848, Hydroxypiracetam, ISF 2522, Neuractiv, Neuromet, Oxiracetam
IUPAC Name 2-(4-hydroxy-2-oxopyrrolidin-1-yl)acetamide
SMILES NC(=O)CN1CC(O)CC1=O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C6H10N2O3/c7-5(10)3-8-2-4(9)1-6(8)11/h4,9H,1-3H2,(H2,7,10)
Formula Weight 158.0691
Chemical Name or Material Oxiracetam
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.