Learn More
Thermo Scientific™ Owl™ S3S Aluminum-Backed Sequencer System Replacement Parts
Click to view available options
Description:
Buffer drain bottle
Replacement Knob (Pack of 6)
Side Clamps; 1 pair
Unit Size:
Each
Pack of 6
Description
Achieve optimal manual sequencing with Thermo Scientific™ Owl™ S3S Aluminum-Backed Sequencer System Replacement Parts, for use with the Owl™ S3S Aluminum-Backed Sequencer System.
Choose from these replacement parts for optimal manual sequencing.
Specifications
Specifications
| Description | Buffer drain bottle |
| For Use With (Equipment) | Owl™ S3S Aluminum-Backed Sequencer System |
| Product Type | Drain Bottle |