Learn More
Thermo Scientific™ Owl™ D4 Horizontal Electrophoresis System Replacement Parts
Click to view available options
Type:
Caster
Gasket
Gel Tray
Unit Size:
Each
Pack of 2
Description
Replace your UVT Gel Tray, gaskets and external caster with Thermo Scientific™ Owl™ D4 Horizontal Electrophoresis System Replacement Parts, for use with the Owl D4 System.
- System design allows you to run one gel tray (51 samples) or stack two gel trays (102 samples) in the buffer chamber while saving valuable bench space.
- Each of the (2) U.V. Transmissible (UVT) gel trays, 16 x 17 cm (W x L), accommodates up to three combs, allowing the user to run up to three series of samples of equal distances.
Specifications
Specifications
| For Use With (Equipment) | Owl™ D4 Electrophoresis System |
| Product Line | Owl |
| Type | Gasket |
| Includes | Gasket Pair |
| Length (Metric) | 17 cm |
| Width (Metric) | 16 cm |