Learn More
Thermo Scientific™ Owl™ D2 Wide-Gel System Replacement Parts
Product Code. 12414607 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Caster
Gasket
Gel Tray
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
12414607 Caster Each
12424607 Gasket Each
12464607 Gel Tray Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12414607 Supplier Thermo Scientific™ Supplier No. D2CST
Only null left
Add to basket
Add to basket
Replace the UVT gel tray, gaskets and external caster with Thermo Scientific™ Owl™ Wide-Gel System Replacement Parts, for use with Owl™ D2 Wide-Gel System.
Replace the UVT gel tray, gaskets and external caster with Thermo Scientific™ Owl™ D2 Wide-Gel System Replacement Parts, for use with Owl™ D2 Wide-Gel System.
  • UVT gel tray incorporates gasketed end gates which provide a leakproof seal without tape
  • Comb slots are positioned so you can arrange up to 12 sample sets on the same gel
TRUSTED_SUSTAINABILITY
Specifications
Description External caster (no trays/combs)
For Use With (Equipment) Owl™ D2 Wide Gel Electrophoresis System
Product Line Owl
Type Caster