missing translation for 'onlineSavingsMsg'
Learn More

OTSSP167 hydrochloride, MedChemExpress

Product Code. p-200095939
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16462677 50 mg 50mg
16449157 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16462677 Supplier Medchem Express Supplier No. HY15512A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress OTSSP167 (OTS167) hydrochloride is a highly potent and ATP-competitive MELK inhibitor with IC50 value of 0.41 nM.

OTSSP167 (OTS167) hydrochloride is a highly potent and ATP-competitive MELK inhibitor with IC50 value of 0.41 nM.

Specifications

Chemical Name or Material OTSSP167 hydrochloride
CAS 1431698-10-0
Color Gray
Purity Grade Notes Research
Molecular Formula C25H29Cl3N4O2
Quantity 50 mg
Synonym OTS167 hydrochloride
Solubility Information DMSO : 33.33 mg/mL (63.62 mM; Need ultrasonic) ∣H2O : 7.14 mg/mL (13.63 mM; Need ultrasonic)
SMILES ClC1=CC(C2=CC=C(N=CC(C(C)=O)=C3N[C@H]4CC[C@H](CN(C)C)CC4)C3=N2)=CC(Cl)=C1O.[H]Cl
Molecular Weight (g/mol) 523.88
Formula Weight 523.88
Percent Purity 99.37%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.