Learn More
Osimertinib, MedChemExpress
Product Code. 16481196
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
500 mg
1 g
Unit Size:
100mg
10mg
1g
200mg
500mg
50mg
5mg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16481196 200 mg 200mg
16476926 100 mg 100mg
16472887 50 mg 50mg
16456818 10 mg 10mg
16409367 5 mg 5mg
16401559 1 g 1g
16441786 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 16481196 Supplier Medchem Express Supplier No. HY15772
MedChemExpress Osimertinib (AZD9291) is a covalent, orally active, irreversible, and mutant-selective EGFR inhibitor with an apparent IC50 of 12 nM against L858R and 1 nM against L858R/T790M, respectively. Osimertinib overcomes T790M-mediated resistance to EGFR inhibitors in lung cancer.
Osimertinib (AZD9291) is a covalent, orally active, irreversible, and mutant-selective EGFR inhibitor with an apparent IC50 of 12 nM against L858R and 1 nM against L858R/T790M, respectively. Osimertinib overcomes T790M-mediated resistance to EGFR inhibitors in lung cancer.
Specifications
Chemical Name or Material Osimertinib
CAS 1421373-65-0
Color Light Brown
Purity Grade Notes Research
Molecular Formula C28H33N7O2
Quantity 200 mg
Synonym AZD-9291 Mereletinib
Solubility Information DMSO : 62.5 mg/mL (125.10 mM; Need ultrasonic)
SMILES C=CC(NC1=CC(NC2=NC=CC(C3=CN(C)C4=C3C=CC=C4)=N2)=C(OC)C=C1N(CCN(C)C)C)=O
Molecular Weight (g/mol) 499.61
Formula Weight 499.61
Percent Purity 99.51%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease