missing translation for 'onlineSavingsMsg'
Learn More

Oseltamivir phosphate, MedChemExpress

Product Code. p-200096303
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16407236

Brand: Medchem Express HY17016

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Oseltamivir phosphate (GS 4104) is a neuraminidase inhibitor recommended for the treatment and prophylaxis of influenza A and B.

Oseltamivir phosphate (GS 4104) is a neuraminidase inhibitor recommended for the treatment and prophylaxis of influenza A and B.

Specifications

Chemical Name or Material Oseltamivir phosphate
CAS 204255-11-8
Color White
Purity Grade Notes Research
Molecular Formula C16H31N2O8P
Quantity 100 mg
Synonym GS 4104
Solubility Information H2O : 100 mg/mL (243.66 mM; Need ultrasonic) ∣DMSO : 100 mg/mL (243.66 mM; Need ultrasonic)
SMILES O=C(OCC)C1=C[C@H]([C@@H]([C@H](C1)N)NC(C)=O)OC(CC)CC.OP(O)(O)=O
Molecular Weight (g/mol) 410.4
Formula Weight 410.4
Percent Purity 98.18%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.