missing translation for 'onlineSavingsMsg'
Learn More

Oseltamivir acid, MedChemExpress

Product Code. 16471807
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16471807

Brand: Medchem Express HY13318

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Oseltamivir acid (GS 4071), the active metabolite of Oseltamivir phosphate, is an orally bioavailable, potent and selective inhibitor of influenza virus neuraminidase (IC50=2 nM) with activity against both influenza A and B viruses.

Oseltamivir acid (GS 4071), the active metabolite of Oseltamivir phosphate, is an orally bioavailable, potent and selective inhibitor of influenza virus neuraminidase (IC50=2 nM) with activity against both influenza A and B viruses.

Specifications

Chemical Name or Material Oseltamivir acid
CAS 187227-45-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C14H24N2O4
Quantity 50 mg
Synonym GS 4071 Ro 64-0802 Oseltamivir carboxylate
Solubility Information DMSO : ≥ 230 mg/mL (808.86 mM) ∣H2O : 125 mg/mL (439.60 mM; Need ultrasonic)
SMILES O=C(O)C1=C[C@H]([C@@H]([C@H](C1)N)NC(C)=O)OC(CC)CC
Molecular Weight (g/mol) 284.35
Formula Weight 284.35
Percent Purity 95.02%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.