Learn More
Thermo Scientific™ Orion™ Electrode Adapter Cables
Click to view available options
Connector Types:
Screw Cap
Meter Input:
2 mm Pin Tip
BNC
Unit Size:
Each
Description
Convert an electrode with one connector to the required connector of another meter by using the Thermo Scientific™ Orion™ Electrode Adapter Cables.
- For use with meters that have BNC, US standard, certain LEMO, British, E DIN, Radiometer, 2mm reference pins or just a stripped lead inputs
- Electrodes must have a US standard, screw cap, Karl Fisher, or BNC connectors
Specifications
Specifications
| Connector Types | Screw Cap |
| Description | 2mm Pin Tip; Screw Cap; Detachable Cable to 2.5 mm Pin Tip Connector |
| For Use With (Equipment) | Meters with 2 mm Reference Pins Connectors, Meters with BNC Connectors, Meters with British Connectors, Meters with Certain LEMO Connectors, Meters with E DIN Connectors, Meters with Radiometer Connectors, Meters with Stripped Lead Inputs Connectors, Meters with US Standard Connectors |
| Type | Electrode Adapter Cable |
| Meter Input | 2 mm Pin Tip |