Learn More
BD Oral Syringes
Click to view available options
Volume (Metric):
1 mL
3 mL
10 mL
Color:
Amber
Unit Size:
Pack of 500
Description
Oral Syringes
Specifications
Specifications
| Color | Amber |
| Latex Free | Yes |
| Product Type | Oral Syringe |
| Volume (Metric) | 10 mL |
| Sterility | Non-sterile |