missing translation for 'onlineSavingsMsg'
Learn More

Optovin, Tocris Bioscience™

Product Code. 30001622
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30001622

Brand: Tocris Bioscience 4901/50

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reversible photoactive TRPA1 activator

Reversible photoactive TRPA1 activator. Stimulates human TRPA1 channels in vitro and enables repeated photoactivation of motor behaviors in wild-type zebrafish (EC50 = 2 μM) and mice, in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 348575-88-2
Molecular Formula C15H13N3OS2
Molecular Weight (g/mol) 315.409
InChI Key YISGMOZSGOGYOU-UHFFFAOYSA-N
Synonym optovin, cbmicro_018200, 5e-5-2,5-dimethyl-1-pyridin-3-ylpyrrol-3-yl methylidene-2-sulfanylidene-1,3-thiazolidin-4-one, 5-2,5-dimethyl-1-pyridin-3-ylpyrrol-3-yl methylidene-2-sulfanylidene-1,3-thiazolidin-4-one
PubChem CID 2890284
IUPAC Name 5-[(2,5-dimethyl-1-pyridin-3-ylpyrrol-3-yl)methylidene]-2-sulfanylidene-1,3-thiazolidin-4-one
SMILES CC1=CC(=C(N1C2=CN=CC=C2)C)C=C3C(=O)NC(=S)S3

Specifications

Quantity 50 mg
Formula Weight 315.41
Percent Purity >99%
Chemical Name or Material Optovin
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.