missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Optovin, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Reversible photoactive TRPA1 activator. Stimulates human TRPA1 channels in vitro and enables repeated photoactivation of motor behaviors in wild-type zebrafish (EC50 = 2 μM) and mice, in vivo.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 315.41 |
| Percent Purity | >99% |
| Chemical Name or Material | Optovin |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction