missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ONX-0914, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
ONX-0914 (PR-957) is a selective inhibitor of low-molecular mass polypeptide-7 (LMP7), the chymotrypsin-like subunit of the immunoproteasome. ONX-0914 blocks cytokine production and attenuates progression of experimental arthritis. ONX-0914 is a noncompetitive irreversible inhibitor of the mycobacterial proteasome (Ki=5.2 μM). ONX-0914 reactivates latent HIV-1 through p-TEFb activation mediated by HSF-1.
Specifications
Specifications
| Chemical Name or Material | ONX-0914 |
| CAS | 960374-59-8 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C31H40N4O7 |
| Quantity | 50 mg |
| Synonym | PR-957 |
| Solubility Information | DMSO : ≥ 35 mg/mL (60.28 mM) |
| SMILES | O=C(N[C@@H](C)C(N[C@@H](CC1=CC=C(OC)C=C1)C(N[C@@H](CC2=CC=CC=C2)C([C@]3(C)OC3)=O)=O)=O)CN4CCOCC4 |
| Molecular Weight (g/mol) | 580.67 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction