missing translation for 'onlineSavingsMsg'
Learn More

Ondansetron hydrochloride dihydrate, MedChemExpress

Product Code. p-200096227
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
1 g
5 g
Unit Size:
100mg
1g
50mg
5g
This item is not returnable. View return policy

Product Code. 16498046

Brand: Medchem Express HYB0002A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ondansetron hydrochloride dihydrate (GR 38032 hydrochloride dihydrate; SN 307 hydrochloride dihydrate) is a serotonin 5-HT3 receptor antagonist used mainly as anantiemetic (to treat nausea and vomiting), often following chemotherapy.

Ondansetron hydrochloride dihydrate (GR 38032 hydrochloride dihydrate

Specifications

Chemical Name or Material Ondansetron hydrochloride dihydrate
CAS 103639-04-9
Color White
Purity Grade Notes Research
Molecular Formula C18H24ClN3O3
Quantity 100 mg
Synonym GR 38032 hydrochloride dihydrateSN 307 hydrochloride dihydrate
Solubility Information DMSO : 100 mg/mL (273.34 mM; Need ultrasonic) ∣H2O : 16.67 mg/mL (45.57 mM; Need ultrasonic)
SMILES O=C1C(CN2C=CN=C2C)CCC(N3C)=C1C4=C3C=CC=C4.[H]Cl.O.O
Molecular Weight (g/mol) 365.85
Formula Weight 365.85
Percent Purity 98.48%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.