Learn More
Ondansetron hydrochloride dihydrate, MedChemExpress
Product Code. 16440758
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
1 g
5 g
Unit Size:
100mg
1g
50mg
5g
Product Code. 16440758
Brand: Medchem Express HYB0002A
MedChemExpress Ondansetron hydrochloride dihydrate (GR 38032 hydrochloride dihydrate; SN 307 hydrochloride dihydrate) is a serotonin 5-HT3 receptor antagonist used mainly as anantiemetic (to treat nausea and vomiting), often following chemotherapy.
Ondansetron hydrochloride dihydrate (GR 38032 hydrochloride dihydrate
Specifications
Chemical Name or Material Ondansetron hydrochloride dihydrate
CAS 103639-04-9
Color White
Purity Grade Notes Research
Molecular Formula C18H24ClN3O3
Quantity 5 g
Synonym GR 38032 hydrochloride dihydrateSN 307 hydrochloride dihydrate
Solubility Information DMSO : 100 mg/mL (273.34 mM; Need ultrasonic) ∣H2O : 16.67 mg/mL (45.57 mM; Need ultrasonic)
SMILES O=C1C(CN2C=CN=C2C)CCC(N3C)=C1C4=C3C=CC=C4.[H]Cl.O.O
Molecular Weight (g/mol) 365.85
Formula Weight 365.85
Percent Purity 98.48%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation